Learn More
Perkin Elmer US LLC 3-Slot Demountable Quartz Torch for Optima 4300/5300/7300 V
Supplier: Perkin Elmer US LLC N0771500
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for use in organic analysis. It is included in the Quick-Change Torch Module for HF Analysis (N0770911), the Organics Sample Introduction Kit (N0770732), and the Quick-Change Torch Module for Oil Analysis (N0770910) for Optima 4300V/5300V/7300V.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.