Learn More
Thomas Scientific Cell Strainer, 40 um, Sterile, Blue, 50/pk
SDP
Supplier:  Thomas Scientific C4040
Encompass
Individually Wrapped, with 5 reducing adapters for use with 15mL tubes. Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh.
Catalog No. NC1304828
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.