Learn More
Waters Corp Tubing Assembly, 40 µm Capillary, Auxiliary Solvent Manager to Injector Valve
Supplier: Waters Corp 430001576
The 40um Capillary from the ASM to the inject valve fits into the 10,000psi nanoACQUITY system.