Learn More
CELLTREAT Scientific Products 40m Cell Strainer, Bulk Packed, Sterile
Supplier: CELLTREAT Scientific Products 229482
CELLTREAT Scientific Products 40¿m Cell Strainer. CELLTREAT Cell Strainers can be used for many applications including isolating cells and removing clumps or debris. Polypropylene housing with nylon mesh filter eliminated the need to fabricate filters from gauze, paper, nylon mesh, or other materials. Extended lip on housing enables aseptic handling with forceps. Elongated gripping tab on housing for easy handling. Cell strainers are designed to universally fit most brands of 50mL conical tubes. Blue color for easy identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile,bulk packed, 10/bulk packed/bag, 50/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.