Learn More
Chemglass Life Sciences Sleeve, PTFE, Medium Length, 40/35 Joint Size
Supplier: Chemglass Life Sciences CG11609
Sleeve, PTFE, Medium Length, 40/35 Joint Size. Reusable .005in thick PTFE sleeve for standard taper joints provides a grease-free seal.