Learn More
Chemglass Life Sciences 4in Diameter X 1/16in Thick Quartz Disc
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CGQ060013
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
4in Diameter X 1/16in Thick Quartz Disc. Commercial quality ground and polished clear fused quartz discs. Edges have a ground finished on all sizes.SIZES OTHER THAN THOSE LISTED ARE AVAILABLE. PLEASE CALL FOR INFORMATION.