Learn More
Selleck Chemical LLC 4-Thiouridine-5mg
SDP
Supplier:  Selleck Chemical LLC E1292
Encompass_Preferred
4-thiouridine (4-Thio-UTP), a thiouridine and a nucleoside analogue, has a role as an affinity label and an antimetabolite.
Catalog No. 50-238-9778