Learn More
Indofine Chemical 4-Hydroxy-3-Methoxyb, 100 Gm
Supplier: Indofine Chemical 01909 100 GM
Molecular Formula: C8H8O4, Molecular Weight: 168.15, Melting Point 209-212C, Appearance: Creamy-white to pale cream fine powder, Storage conditions: Store in a cool, dry place