Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(4-Dimethylaminophenyl)di-tert-butylphosphine, TCI America™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
$111.00 - $295.00
Specifications
CAS | 932710-63-9 |
---|---|
Melting Point | 69°C |
Color | White-Yellow |
Molecular Formula | C16H28NP |
MDL Number | MFCD09265102 |
Specifications
932710-63-9 | |
White-Yellow | |
MFCD09265102 | |
IQTHEAQKKVAXGV-UHFFFAOYSA-N | |
4-ditert-butylphosphanyl-N,N-dimethylaniline | |
11714598 | |
Crystalline Powder |
69°C | |
C16H28NP | |
4-(Di-tert-butylphosphino)-N,N-dimethylaniline, APhos, A-taPhos, Amphos | |
CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C | |
265.381 | |
265.38 |
Safety and Handling
TSCA : No
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title