Learn More
Electron Microscopy Sciences 4 Compartment Tissue Basket with Lid, 15 mm diameter, 10 mm height
Supplier: Electron Microscopy Sciences 7019428
EMS has introduced and is now available a variety of Stainless steel baskets with lids and multiple compartments for critical point Drying. These baskets are either 15 mm in diameter or 23 mm.
Diameter: 15 mm
Height: 10 mm