Learn More
Chemglass Life Sciences 3-Port Harvest Tube Assembly for 3L VSA Vessels, Complete
Supplier: Chemglass Life Sciences CLS143770
3-Port Harvest Tube Assembly for 3L VSA Vessels, Complete. Our 3-port harvest assemblies are designed for insertion into our vertical sidearm bioreactor vessels. They are manufactured with 1/4” and 3/8” 316L stainless steel tubing and facilitate rapid removal of liquid from bioreactor vessels. Harvest tube assemblies have an electro-polished finish of Ra 20µ or better, inside and out. Includes a blue open top GL45 polypropylene cap with a white silicone gasket.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.