Learn More
Waters Corp Flowcell Rebuild Kit, 3 mm, 2487
Supplier:  Waters Corp WAS081347
Encompass_Preferred
The 3mm Flow Cell Rebuild Kit contains (1) entrance lens screw, (1) entrance lens holder, (1) entrance lens (curved), (10) gaskets, (1) exit window (flat), (1) exit window holder, (1) exit window screw, (1) cuvette lens screw, (1) cuvette lens, and (1) split ring (cuvette lens holder).
Catalog No. 50-793-467