Learn More
3M™ Virtua™ Protective Eyewear
Click to view available options
Clear color; Clear temple; Clear lens; Hard coat; Frameless; Standard size
Description
- Economy-rated glasses merge style, fit, lightweight comfort, and affordability
- Lightweight design weighs less than an ounce for comfort
- Wraparound design provides extended eye protection and unobstructed viewing
- Unisex styling fits a variety of facial profiles
- Polycarbonate lenses absorb 99.9% UVA and UVB
- 3M Catalog# 11326-00000-20
Specifications
Product Type | Protective eyewear |
Model | 11326-00000-20 |
Lens Material | Polycarbonate |