Learn More
3M™ Qualitative Fit-Test Kit, Sweet
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter.
Supplier: 3M™ 7100046674
Includes: Laminated instruction booklet (multilingual); .9 oz. (55ml) sensitivity solution; 1.9 oz. (55mL) fit test solution; Two nebulizers; Hood; Collar assembly; Two sets replacement nebulizer inserts. Optional training and fit test case includes fit-test kit materials as well as training materials (videos, posters and literature).
Description
- Works by spraying a sweet solution into the hood environment
- If wearer does not detect sweet taste, the fit test has been passed
- 3M Catalog# FT-10, FT-11, FT-12, FT-20
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
Individual 1.9 oz. (55mL) bottles of sensitivity solution or fit test solution also available.
Compliance
Meets the performance criteria for fit testing respirators under OSHA 29 CFR 1910.134, Appendix A.
Specifications
Training and Fit Test Case | |
Qualitative fit test kit | |
Laminated instruction booklet, sensitivity solution, fit test solution, two nebulizers, hood, collar assembly, two sets replacement nebulizer inserts, premium case and training materials: videos, posters, and literature | |
FT-20 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.