Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ PELTOR™ Wireless Communication X Series Ear Muff Accessory
Click to view available options
X Series Ear Muff Accessory
Description
- Easily attaches to 3M PELTOR X Series Ear Muffs
- Designed for communicating in noisy environments
- Noise-canceling microphone
- Bluetooth wireless technology for connecting to smart phones
- Resistant to harsh environmental elements including temperature fluctuations, sweat, and rain
- Comfortable for all day wearability
- Talk/Streaming Time: Approx. 8 hours
- Standby Time: Approx. 250 hours
- Charge Time: Approx. 3 hours
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Black |
ANSI S3.1974 | Yes |
For Use With (Equipment) | X Series Ear Muffs |
For Use With (Application) | With Bluetooth™ technology, allows workers to stay connected to smart phone. |
Model | WS-CUSH |
Product Type | Accessory for X Series Ear Muff |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction