Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Audio Input Cable

Click to view available options
Model:
FL6H
FL6M
FL6N
Product Type:
Audio Input Cable 2.5 mm mono plug
Audio Input Cable 2.5 mm stereo plug
Audio Input Cable 3.5 mm mono plug

Patch cable that connects certain PELTOR™ headsets to microphones, radios or other personal entertainment devices (listen only). Includes a 36-inch straight cable.

  • Available plug types include 2.5 mm mono, 3.5 mm mono and 2.5 mm stereo
  • For use with PELTOR™ headsets that have an available J22 auxiliary port
  • Flexible, insulated connection cords made from durable polyurethane
  • Meets UL specifications
  • Mono plugs also connect to VHF/UFH radios.
  • Includes 36-inch straight cable

Compliance

3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

For Use With (Equipment) PELTOR™ HT Series Listen-Only Headsets to Connect to Speaker Microphones
Model FL6N
Product Type Audio Input Cable 2.5 mm stereo plug
Material Polyurethane
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit