Learn More
3M™ Noise Indicator NI-100
Alerts users to potentially dangerous noise levels
Supplier: 3M™ NI100
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
- Small size and lightweight design; just clip to a shirt or jacket
- LED light indicates when noise levels exceed a potentially hazardous threshold
- Rechargeable battery operates for up to 200 hours between charges
Red Flashing LED: Above 85dBA (hearing protection may be required)
Green Flashing LED: Below 85dBA (hearing protection may not be necessary)
Operating/Storage Temperature: -10 to +50°C (14 to 122°F)
Humidity Range: 0 to 95%; noncondensing
Accuracy: 3dB
Alert Level: Flashes red when >85 dBA / Flashes green when <85dBA
Battery Type: Li-Poly (flat cell); recharge after ≈200 hours
Battery Life: Approximately two years of charging cycles
Charging: Mini-USB cable plugs into bottom port of device (sold separately; contact your Fisher Safety Customer Service Representative)
Auto-Power Off: At 10 hours
Dimensions (L x W x D): 2 x 1.4 x 0.5 in. (5.1 x 3.6 x 1.3cm)
Order Info
USB cable charger (Cat. No. 17-979-000) sold separately.
Specifications
OSHA 1910.05 | |
1.27 cm | |
Noise Indicator | |
To measure potentially hazardous noise levels | |
2 in. | |
±3 dB | |
57.2°C to 251.6°C | |
17 g | |
3.55 cm |
0.5 in. | |
2 x 1.4 in. (5.08 x 3.55 cm) | |
LED | |
0% to 95% (Non-Condensing) | |
5.08 cm | |
14°F to 122°F | |
0.6 oz. | |
1.4 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.