Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ N95 and P95 Particulate Filters (5000/6000 Series)
NIOSH approved for environments containing certain non-oil based particles. Use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501.
Supplier: 3M™ 7000002054
Requires : Filter retainer 17-673B for use with 5000 Series Respirators and 6000 Series Cartridges.
Description
The 3M™ Particulate Filter 5N11, N95 helps provide respiratory protection in environments containing certain non-oil based particles. When properly fitted, use with appropriate cartridges for various applications including spray painting, petrochemical and chemical manufacturing, and pesticide spraying for concentrations up to 10 times the Permissible Exposure Limit (PEL) with half facepieces, or 50 times PEL with quantitatively fit tested full facepieces. Filter protects against: 1) solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances; 2) liquids or oil based particles from sprays that do not emit harmful vapors; and 3) metal fumes produced from welding, brazing, cutting, and other operations involving heating of metals.NIOSH (National Institute for Occupational Safety and Health) approved for environments containing certain non-oil based particles. This filter has been rated N95, which means it filters at least 95% of airborne particles, but is not resistant to oil.Breathing hazardous particles can pose a risk to your health. NIOSH, a Federal government regulatory agency, has tested and approved the 3M™ Particulate Filter 5N11 to help reduce exposure to certain particles.- NIOSH approved for environments containing certain non-oil-based particles
- Replaceable particle filter for cost savings
- Wide range of applications reduces inventory needs and training requirements
- Designed for use with 3M™ Respirators 5000 Series or 3M™ Cartridges 6000 Series or 3M™ Filter Adapter 603, and 3M™ Filter Retainer 501
- 3M Catalog# 5N11 , 3M Catalog# 5P71
Specifications
Filter Cartridge | |
P95 Particulate Filter | |
3M™ respirator 5000 series or 3M™ cartridge 6000 series or 3M™ filter adapter 603 and 3M™ filter retainer 501 | |
4.2 in. | |
3.4 in. |
NIOSH | |
Cleaning, demolition, facility maintenance, grinding, painting, sanding, welding | |
5P71 | |
10.6 cm | |
8.6 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction