Learn More
3M™ E-A-Rfit™ Dual-Ear Enclosure Assembly Speaker
Click to view available options
E-A-Rfit™ Dual-Ear Enclosure Assembly Speaker
Description
- For use with E-A-Rfit™ dual-ear validation system
- Connects to an AC power source
- Compact and easy to set up
- Presents a test signal across seven test frequencies (125 Hz, 250 Hz, 500 Hz, 1000 Hz, 2000 Hz, 4000 Hz, 8000 Hz) in under five seconds
- USB Cable connects speaker to computer (not included)
- Dual-element microphone (Catalog No. 19-122-402) plugs into back of speaker (not included)
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
For Use With (Equipment) | E-A-Rfit™ Dual-Ear Validation System |
Model | 073-783 |
Product Type | E-A-Rfit™ Dual-Ear Enclosure Assembly Speaker |