Learn More
3M™ E-A-R™ Express™ Corded Metal Detectable Ear Plugs
Click to view available options
Classic, Metal Detectable, Corded
Description
- Designed with detectable metal components for food processing and other industries
- Low-pressure in the ear canal for enhanced comfort
- Poly bag keeps each pair of ear plugs clean before use
- Moisture-resistant helps maintain a consistent expansion time, even in humid situations
- Slightly longer than regular size 3M™ E-A-R™ Classic™ ear plugs for higher noise reduction
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 | Yes |
Cord Color | Blue |
CSA Z94.2 | Class AL |
Size | Large |
For Use With (Application) | Food Processing, Manufacturing, Pulp and Paper, Pharmaceutical, Tobacco Processing Industries and Other Various Industries That Require Detectable Products to Prevent Contamination |
High Visibility | Yes |
Model | 311-4101 |
Packaging Type | Poly bag |
Pairs per Package | 200 Pairs/Pk. |
Plug Material | Foam |
Show More |