Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Disposable Protective Oversleeve 444
Click to view available options
White
Description
Helps protect arms against light liquid splashes and hazardous dusts
- Tapered design and elastic around top and bottom of sleeve for secure and easy fit
- Anti-static treated
- 3M Catalog# 444
Specifications
Size | Universal |
Construction | Microporous laminate |
Material | Microporous Fabric |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic |
Model | 444 |
For Use With (Application) | General maintenance, electronics manufacturing, catering, clean rooms, beauty industries, hygiene, laboratories |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction