Learn More
Supply Solutions 3M - Petrifilm Count Plate. Type: Count Plate. Incubation: 24 Hours; For Use With: Aerobic Count.; Type: Count Plate; Incubation: 24 Hours
Supplier:  Supply Solutions 3M 6400
Encompass_Preferred
In today's environment of heightened focus on food safety and increasingly stringent quality requirements, labs are under pressure to provide tests that are fast, reliable and consistently accurate. 3M Petrifilm Plates have demonstrated consistent performance that has been cited globally with more than 200 certificates, recognitions, validations, and peer-review publications. For more than 30 years, food safety professionals around the world have put their trust in 3M Petrifilm Plates. That's because 3M Petrifilm Plates have streamlined, standardized and simplified the process of microbial quantitative indicator testing, improving productivity and helping ensure the highest levels of product quality.; Type: Count Plate; Incubation: 24 hours; Min. Population: -; Shelf Life: 1 year after open; For Use With: Aerobic Count; Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes; 3M Petrifilm Plates use 75% less energy, use nearly 80% less w
Catalog No. NC0160490
Explore available promotions