Learn More
Ace Glass, Inc. 3000mL jacketed filter reactor, complete with 28/15 ballpoint inlet/outlets, 150mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings
Supplier: Ace Glass, Inc. 638660
3000mL jacketed filter reactor, complete with 28/15 ballpoint inlet/outlets, 150mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings