Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Wave Demonstration Machine, Complete Set
SDPSCI_ED

Supplement to demonstration wave machine comprising module with short pendulum bars, one transition module and two module couplers.

Supplier:  3B Scientific™ U45011

Catalog No. S08157


Add to Cart

Description

Description

If the two models with differing bar lengths and thus differing wave velocities are coupled together, then reflections can be observed at the point where they are joined together. This can be avoided by adding the transition module.

  • Length of bar module 1: 46cm
  • Length of bar module 2: 23cm
  • Length of bars for transition module: 23 to 46cm
  • Total length: 244cm

Specifications

Specifications

Wave demonstration machine, complete set
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit