Learn More
3B Scientific™ Interferometer
Permits generation of clear and well-defined interference patterns in daylight.
Supplier: 3B Scientific™ U10350
Description
- Comprises high-grade optical components mounted on heavy, rigid base plate for precise and reproducible measurements
- Reflective observation screen has adjustable inclination
- Predefined component positions allow quick rearrangement to ensure extremely rapid preparation for various experiments
- Stable plastic box for storing mounted and adjusted interferometer
- Michelson interferometer
- Fabry-Perot interferometer
- Determination of refractive index of glass
- Determination of refractive index of air
- Twyman-Green test for optical components (qualitative)
- Beam splitter: diameter 40mm, evenness λ/10 (front side), λ/4 (rear side)
- Surface-coated mirror: dimensions 40 x 40mm2, evenness <λ/2, eccentric reduction ∽1:1000 (individual calibration specified on eccentric base)
- Base plate: weight 5.5kg, dimensions: 245 x 330 x 25mm3
Specifications
Interferometer |