Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Gynecologic Simulator
Represents female adult lower body with anatomically accurate female genital organs, fimbriae and ovaries.
Supplier: 3B Scientific™ W45043
Includes: 10 fallopian tubes, talcum powder and carrying bag, normal and abnormal cervices as well as normal and pregnant uteri
Description
- For training of various pelvic examinations, laparoscopy, mini-laparotomy as well as recognition of pathologies and placing of contraceptive devices
- Opened model to make inner structures visible
- supplied with closed abdomen
- Vaginal speculum and bi-manual pelvic examination
- Uterine sounding
- Insertion and removal of contraceptive devices such as IUD, 75mm diaphragm, or female condom
- Laparoscopic visualization and occlusion of fallopian tubes
- Mini-laparotomy
- Supplied with normal and abnormal cervices as well as normal and pregnant uteri which can be easily changed:
- One anteverted and one retroverted parous uterus
- Two pregnant uteri (ten-week/twenty-week)
- Five normal cervices with patent as for IUD insertion/removal
- Four cervices with pathologies
Specifications
20.1 x 18.1 x 10.0 in. | |
Training of various pelvic examinations including laparoscopy, mini-laparotomy as well as recognition of pathologies and placing of contraceptive devices | |
Talcum powder, Soft carrying bag | |
16.70 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction