Learn More
3B Scientific™ Catheterization Trainers
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Material gives realistic feel and flexibility.
Supplier: 3B Scientific™ W44789M
Description
- For training of urinary catheterization
- Utilizes 16 FR catheter and can be used to demonstrate balloon inflation procedures
- Fluid reservoir allows for urine output upon successful completion of inertion procedure
- Instruction for to teach sterile technique and insertion and removal of catheter, as well as practice preparing patient for procedure
Specifications
Trainer, Male Catheterization | |
Ideal for urinary catheterization | |
1.13 kg |
35.6 x 22.9 x 12.7 cm (14.0 x 9.0 x 5.0 in.) | |
2.49 lb. |