Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 380 Foam Nitrile Coated Seamless Knit Nylon Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Long-lasting grip in both wet and dry environments
- Ensures tactile precision while working with small oily components or lifting heavy objects
- Elasticated wrist cuff prevents debris from entering the glove
- Open-back design allows air and moisture to escape
Specifications
Construction | Seamless Knit |
Coating Material | Nitrile foam |
Color | Blue |
Hand | Specific |
Latex Free | No |
Material | Nylon |
Product Type | Seamless Knit Gloves |
Size | Small |
For Use With (Application) | Automotive Repairs, and Intricate/Oily Parts Handling |
Glove Cuff Style | Knit |
Show More |