Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ 37 Piece Lab Starter Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Click to view available options
37 Piece Lab Starter Kit; Material: Borosilicate Glass
Description
Kit includes:
- 3 x 50 mL Low Form Borosilicate Glass Graduated Beaker with Spout
- 3 x 100 mL Low Form Borosilicate Glass Graduated Beaker with Spout
- 6 x 250 mL Low Form Borosilicate Glass Graduated Beaker with Spout
- 2 x 100 mL Graduated Borosilicate Glass Reagent Bottles with Screw Cap
- 3 x 250 mL Graduated Borosilicate Glass Reagent Bottles with Screw Cap
- 3 x 500 mL Graduated Borosilicate Glass Reagent Bottles with Screw Cap
- 2 x 1000 mL Graduated Borosilicate Glass Reagent Bottles with Screw Cap
- 1 x 100 mL Class B Borosilicate Glass Graduated Cylinder with Spout, Blue Plastic Guard and Hex Base
- 1 x 250 mL Class B Borosilicate Glass Graduated Cylinder with Spout, Blue Plastic Guard and Hex Base
- 1 x 500 mL Class B Borosilicate Glass Graduated Cylinder with Spout, Blue Plastic Guard and Hex Base
- 3 x 250 mL Wide Neck Borosilicate Glass Conical Flask
- 6 x 500 mL Wide Neck Borosilicate Glass Conical Flask
- 3 x 1000 mL Wide Neck Borosilicate Glass Conical Flask
Specifications
Capacity (Metric) | 50 to 1000 mL |
Material | Borosilicate Glass |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Scale | White Graduated Scale |
Style | Low Form |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction