Learn More
MSC 3M 7000058437 3/4" x 66' x 7 mil Purple Vinyl Electrical Tape Rubber Adhesive, 32 to 220°F, 1,250 V/mil, Series 35
Supplier:  MSC 54041496
Encompass
3M 7000058437 3/4" x 66' x 7 mil Purple Vinyl Electrical Tape Rubber Adhesive, 32 to 220°F, 1,250 V/mil, Series 35
Catalog No. NC1682514
Please call customer service for current pricing. \n1-800-766-7000