Learn More
MSC 3M 7000006095 3/4" x 66' x 7 mil Blue Vinyl Electrical Tape Rubber Adhesive, 32 to 220°F, 1,250 V/mil, Series 35
Supplier:  MSC 54041199
Encompass
3M 7000006095 3/4" x 66' x 7 mil Blue Vinyl Electrical Tape Rubber Adhesive, 32 to 220°F, 1,250 V/mil, Series 35
Catalog No. NC1614809
Please call customer service for current pricing. \n1-800-766-7000