Learn More
Cole-Parmer Koflo 1/4-21 316 SS Tube Mixer, 21 Elements; 1/4" OD
Supplier: Cole-Parmer 0466956
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings