Learn More
Chemglass Life Sciences 30L Process Reactor, Complete, Air Motor, 79.0" OAH with Motor, 20.9" Clearance Below Valve
Supplier: Chemglass Life Sciences CG1974630
30L Process Reactor, Complete, Air Motor, 79.0" OAH with Motor, 20.9" Clearance Below Valve.Features:• Motor Mount Arm Can Unlock And Swing Out Giving Greater Access To Reactor• Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings• Electric And XP Motor Has Separate Controller With Digital Display• Reactor Support Plate Holds Reactor Securely And Allows Easy Installation/Removal Of Reactor By One Person• Lower Safety Ring And Lower Mantle Base• Heavy Duty Lockable Casters • Electric Or Air Motor Optional XP Motor Available• Open Frame Design Provides Excellent Accessibility• 7-Neck Lid With Large Powder Addition Port• Ridge Clamp Accepts 1/2” Rod And Adds Additional Support For Large Glassware• #304 S.S. Cooling Coil PTFE Coated • Digital Temperature Controller• Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.