Learn More
Wesco Industrial Products 30BT Standard Series Drum Truck
Supplier: Wesco Industrial Products 240005
Multi-functional drum truck allows you to move drums on four wheels and acts as a drum cradle for storage or dispensing. 16.5" drain height. With two wheels resting on floor, 28" to bottom of lowered chime hook. Steel drums only. Handles 30, 55 & 85. Capacity 1000 lbs. 10" Polyolefins Wheels (front), 6" Polyolefin Wheels (rear). gallon steel drums.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.