Learn More
METTLER TOLEDO 30102935 INPRO 6860I
Supplier:  METTLER TOLEDO 30102935
Encompass
30102935 Inpro 6860i Optical oxygen SensorDimensions: 12mm diameter x 590 mm longOperating range: 8 ppb to SaturationAccuracy: better than +/- 1%Measuring Principle: Fluorescense QuenchingSensor Body Material: 316L SSAutomatic Temperature compensation (5 to 60C)Process Pressure range; 0.2 to 6 Bar (2.9 to 87 psig) *22.03
Catalog No. NC2110249