Learn More
Chemglass Life Sciences Glas-col Mantle Tripod Support for Series inMin, Mantle Size 3000mL, 14in Fixed Height, 259mm ID
Supplier: Chemglass Life Sciences CG1100601
Glas-col Mantle Tripod Support for Series inMin, Mantle Size 3000mL, 14in Fixed Height, 259mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.