Learn More
Purus™ 30-Layer Cleanroom Tacky Mats: PROMO
Remove dirt and dust from foot traffic and equipment wheels
Supplier: Purus™ PM 3636 34 BPM
Description
- 1.4 or 1.8mil low-density polyethylene film layers fully coated with a particle grabbing acrylic adhesive
- Suitable for use in industrial, semiconductor, pharmaceutical, biotech and telecommunications cleanrooms
- Consistent adhesive coverage for maximum performance on each layer
- Numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers
Specifications
36 x 36 in. | |
Polyethylene | |
Adhesive coating |
Blue | |
For use in industrial, semiconductor, pharmaceutical, biotech, and telecommunications cleanrooms | |
1.8 mil |