Learn More
Market Lab 3-Tier Vial and Control Rack
Supplier: Market Lab 13224
14.25"W x 9.125"D x 4.625"H. Unique stadium design increases capacity while saving valuable bench and refrigerator space. The plastic label holder extends the full length of each level for easy vial identification. Adjustable dividers allow for customization and storage flexibility.