Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex 3 Ring Binder Holder/Pen Holder
SDP

Super durable PETG construction.

$87.20 - $266.00

Specifications

Type Binder Holder
Color Clear
Material PETG
Height (Metric) 23 cm
Length (Metric) 8 cm
View More Specs

Products 3
Catalog Number Mfr. No. Mounting Type Price Quantity & Availability  
Catalog Number Mfr. No. Mounting Type Price Quantity & Availability  
22-000-511
SDP
View Documents
Poltex
BINDRPENM
Magnetic
Each for $266.00
Add to Cart
 
22-000-512
SDP
View Documents
Poltex
BINDRPENT
VHB Tape
Each for $87.20
Add to Cart
 
22-000-510
SDP
View Documents
Poltex
BINDRPENW
Wall
Each for $158.60
Add to Cart
 
Description

Description

  • Holds 3 ring binder and pens in side pocket.
  • Mounts with powerful magnets, VHB Tape or screws/anchors to wall.
Specifications

Specifications

Binder Holder
PETG
8 cm
Clear
23 cm
31 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit