Learn More
Chemglass Life Sciences 3-Port Thermowell Assembly for 8L VSA Vessels, Complete
Supplier: Chemglass Life Sciences CLS143762
3-Port Thermowell Assembly for 8L VSA Vessels, Complete. Our 3-port large diameter thermowell assemblies are designed to accommodate a wide variety of RTD’s and dial thermometers of different diameters for measuring the process temperature in our vertical sidearm bioreactor vessels. All 316L stainless steel tubes have an electro-polished finish of a Ra 20µ or better, inside and out. These assemblies feature a thermowell tube with 0.375” OD and 0.305”ID that will accept all 1/4” thermometer probes. These assemblies include a blue open top GL45 polypropylene cap with a white silicone gasket.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.