Learn More
Chemglass Life Sciences Vacuum Manifold, Right Handed, 3 Ports, 0-4mm Chem-Cap Valve, 240mm Overall width
Supplier: Chemglass Life Sciences CG443401
Vacuum Manifold, Right Handed, 3 Ports, 0-4mm Chem-Cap Valve, 240mm Overall width. General purpose single bank vacuum manifold with 3 ports. Main tube is 38mm OD with a serrated hose connection, having the smallest serration 13.5mm and the largest serration 15.5mm, located at either the left or right side for vacuum connection. Ports are 0-4mm Chem-Vac Chem-Cap valves with serrated hose connections. The 0-4mm has the smallest serration 10mm and the largest serration 11mm. Ports are spaced 60mm center to center for the 0-4mm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.