Learn More
Chemglass Life Sciences 3-Port Airfree Schlenk Vacuum Manifold, Single Bank, 315mm OAL
Supplier: Chemglass Life Sciences AF006101
3-Port Airfree Schlenk Vacuum Manifold, Single Bank, 315mm OAL. Single bank vacuum manifold with 0-8mm Chem-Vac™ Chem-Cap valves mounted on a 45 angle to the user permitting easy operation inside of a fume hood. Ports are spaced 75mm center to center. All connection ports are #15 o-ring connections that incorporate a greasefree o-ring seal. Manifold is modular in design allowing the user tremendous flexibility in performing experiments. Adapters from #15 o-ring joints to standard taper components, serrated hose connections or 3/8" Cajon fittings are available to allow the ports to be used with existing glassware. Replacement Chem-Vac™ Chem-Cap plug is listed below.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.