Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-O-tert-Butyldimethylsilyl-4,6-O-(4-methoxybenzylidene)-D-glucal, 97%, Thermo Scientific™

Click to view available options
Quantity:
250 mg
1 g
384346-91-2
C20H30O5Si
378.54
MFCD01863523
ZTRSSQRWGPRALW-UHFFFAOYNA-N
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
71311532
COC1=CC=C(C=C1)C1OCC2OC=CC(O[Si](C)(C)C(C)(C)C)C2O1
384346-91-2
C20H30O5Si
378.54
MFCD01863523
ZTRSSQRWGPRALW-UHFFFAOYNA-N
3-o-tert-butyldimethylsilyl-4,6-o-4-methoxybenzylidene-d-glucal, 4ar,8r,8ar-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl oxy tert-butyl dimethylsilane
71311532
COC1=CC=C(C=C1)C1OCC2OC=CC(O[Si](C)(C)C(C)(C)C)C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Quantity 250 mg
IUPAC Name [(4aR,8R,8aR)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl]oxy-tert-butyl-dimethylsilane
Formula Weight 378.53
Percent Purity 97%
Chemical Name or Material 3-O-tert-Butyldimethylsilyl-4,6-O-(4-methoxybenzylidene)-D-glucal

TSCA : No

Recommended Storage : Keep cold