Learn More
3-(Methylsulfonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Description
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep Cold
Specifications
Melting Point | 90°C to 96°C |
Quantity | 5 g |
Formula Weight | 215.03 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Methylsulfonylamino)benzeneboronic acid |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.