Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, (U.S.P. Test Solution), Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with “TS or Test Solution”, that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
UN Number | UN1230 |
Grade | Chemical Solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction