Learn More
Electron Microscopy Sciences Spot Plate Polypropylene 3 Cavities
Supplier: Electron Microscopy Sciences 7157440
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These spot plates have three depressions 21 mm diameter x 7 mm deep. The tray is 28 mm x 85 mm. Made from low density polyethylene and will withstand temperatures up to 80 degree C.