Learn More
Electron Microscopy Sciences Spot Plate Polypropylene 3 Cavities
SDP
Supplier:  Electron Microscopy Sciences 7157405
Encompass_Preferred
These spot plates have three depressions 21 mm diameter x 7 mm deep. The tray is 28 mm x 85 mm. Made from low density polyethylene and will withstand temperatures up to 80 degree C.
Catalog No. 50-949-148