Learn More
Cole-Parmer Lightnin 809418 316 SS axial flow impeller ( A-100), 3.1" dia x 5/16" bore dia
Supplier: Cole-Parmer 0456054
Axial Flow Impellers (LIGHTNIN A-100) with SUPERPITCH design — 1.5 pitch ratio for greater batch mixing capabilities. Standard axial flow; use for wetting dry powders.