Learn More
BioSpec Products Glass Beads 3.5mm 1 lb
Supplier: BioSpec Products 11079135
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating." Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb