Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3,4-(Methylenedioxy)benzyl isothiocyanate, 94%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.
Solubility
Hydrolyses in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
3,4-(Methylenedioxy)benzyl isothiocyanate is used as intermediates. Used in nucleophilic addition reactions.
Solubility
Hydrolyses in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
Specifications
Density | 1.32 |
Boiling Point | 139°C to 140°C (1 mmHg) |
Odor | Pungent |
Refractive Index | 1.615 |
Quantity | 1 g |
UN Number | UN2922 |
Beilstein | 1428941 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyses in water. |
Formula Weight | 193.23 |
Show More |
Safety and Handling
Hazard Category | H302+H312-H314-H331-H335 |
Hazard Statement | GHS H Statement H331-H314-H318-H302-H312 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction